Learn More
3M™ Particulate Respirator, Series C100
Lightweight and comfortable particulate respirators provide effective protection against dust and mist.
€ 12.10 - € 18.95
Specifications
Material | Polypropylene |
---|---|
Product Type | General Purpose Particulate Respirator |
Sterility | Non-sterile |
FDA Cleared Surgical Mask | No |
Product Code | Brand | Filter Type | With Exhalation Valve | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Type | With Exhalation Valve | Price | Quantity & Availability | ||||||
13587000
|
3M™
7000088840 |
FFP1 | No |
€ 17.50
Pack of 20 |
In Stock
Log in to see stock. ![]() |
||||||
13597000
|
3M™
7000088842 |
FFP2 | No | N/A | N/A | N/A | |||||
13507010
|
3M™
7000088841 |
FFP1 | Yes |
€ 12.10
Pack of 10 |
In Stock
Log in to see stock. |
||||||
13517010
|
3M™
7000088843 |
FFP2 | Yes | N/A | N/A | N/A | |||||
Description
Provides cost-effective, lightweight, comfortable, and effective protection against dust and mist.- The convex shape, twin strap design, nose foam and nose clip ensure comfortable wear over a range of face sizes
- Exhalation valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions
Specifications
Polypropylene | |
General Purpose Particulate Respirator | |
Non-sterile | |
No |
Product Title
3M™ Particulate Respirator, Series C100