missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ LDPE Wide Mouth Bottles without Closures

Designed with a wide opening for easy filling and cleaning. Kautex™ Wide Mouth Bottles are ideal for liquids, pastes and granules.

Brand:  Kautex™ 2000070536

 View more versions of this product

Product Code. 15951391

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Manufactured from low density polyehtylene (LDPE) for flexibility and toughness
  • Natural translucent color, round bottles
  • Ideal for dispensing, sending samples, adhesives, transportation and storage 
  • Closures available separately
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1500 mL
LDPE
No
Round
Wide Neck
1Each
No
Wide Neck Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kautex™ LDPE Wide Mouth Bottles without Closures > Capacity: 1500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.