Learn More
Kautex™ LDPE Wide Mouth Bottles without Closures
Designed with a wide opening for easy filling and cleaning. Kautex™ Wide Mouth Bottles are ideal for liquids, pastes and granules.
Brand: Kautex™ 2000070535
This item is not returnable.
View return policy
Description
- Manufactured from low density polyehtylene (LDPE) for flexibility and toughness
- Natural translucent color, round bottles
- Ideal for dispensing, sending samples, adhesives, transportation and storage
- Closures available separately
Specifications
1000 mL | |
LDPE | |
No | |
Round |
Wide Neck | |
1Each | |
No | |
Wide Neck Bottle |