Learn More
Kappes Systeme™ Longitudinal Divider
Click to view available options
Height (Metric):
105 mm
145 mm
Unit Size:
Each
Specifications
Specifications
| Product Type | Longitudinal Divider |
| Color | Zinc |
| For Use With (Equipment) | Secure and professional storage |
| Quantity | 1/Pk. |
| Height (Metric) | 145 mm |