Learn More
Fisherbrand™ Long-Stem Analytical Funnels
GREENER_CHOICE
Product Code. 15122875 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
12 mL
23 mL
37 mL
50 mL
100 mL
150 mL
225 mL
Height (Metric):
7.5 cm
8.3 cm
10.2 cm
10.9 cm
14.1 cm
16.1 cm
17.7 cm
Length (Metric) Stem:
5cm
5.2cm
6.1cm
6.5cm
8cm
8.8cm
9.9cm
Unit Size:
Each
Pack of 12
Pack of 24
Pack of 36
Pack of 4
Pack of 6
Product Code. 15122875
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Molded polypropylene with external ridges and outside ribs to prevent vapor lock
  • Inside ribbed surface slopes for quick filtration
  • Use standard filter paper
  • Autoclavable to 120°C (248°F)
  • Bowl angle: 58°
Specifications
Diameter (Metric)Top 6.6cm
Capacity (Metric) 50 mL
Material Polypropylene
Height (Metric) 10.9 cm
Angle 58° Bowl
Autoclavable Autoclavable
Diameter (Metric) Filter 11cm
Length (Metric) Stem 6.5cm