Learn More
Endecotts™ Locks
Use these locks with Endecotts™ sieves. Endecotts™ Locks are widely used with test sieves for all types of laboratory sampling and particle size analysis.
Brand: Endecotts™ 200S/STLID
This item is not returnable.
View return policy
Description
Use these locks with Endecotts™ sieves. Endecotts™ Locks are widely used with test sieves for all types of laboratory sampling and particle size analysis.
- Locking assembly for Endecotts Sieves
- ISO 3310-1 Compliant
Specifications
Lock | |
200 mm | |
1 Ea. |
ISO 33101 | |
For sieves |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Endecotts™ Locks > Outer Diameter: 200mm