missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMX1A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5115517
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total LMX1A.
The LIM homeobox transcription factor 1 (LMX1A) belongs to the LIM-homeodomain family. Members of this family are known to be important for pattern formation during development. LMX1A binds an A/T-rich sequence and functions in the nucleus as a transcriptional activator to the insulin gene promoter. In the developing embryo, LMX1A is expressed along the neuraxis and leads to the development of the roof plate of the vertebrae. While LMX1A is crucial for the differentiation of dopamine neurons, the related LMX1B appears to be important for both the development and the survival of these cells. It has been suggested that LMX1A might be a useful biomarker or risk factor in tumor aggressiveness.Specifications
LMX1A | |
Polyclonal | |
Unconjugated | |
Lmx1a | |
dr; dreher; LIM homeobox transcription factor 1 alpha; LIM homeobox transcription factor 1, alpha; LIM homeobox transcription factor 1-alpha; LIM/homeobox protein 1.1; LIM/homeobox protein LMX1A; LMX1; LMX-1; Lmx1.1; LMX-1.1; Lmx1a; MGC87616; shaker short-tail; sst | |
Rabbit | |
Affinity chromatography | |
RUO | |
110648, 289201, 4009 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q8TE12, Q9JKU8 | |
Lmx1a | |
A synthesized peptide derived from human LMX1A(Accession Q8TE12), corresponding to amino acid residues R152-L202. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction