Learn More
Motic™ LM Plan Achromatic Objective
The BA310MET series presents an affordable and powerful microscope line for incident light applications in all material sciences.
Brand: Motic™ 1101001704512
This item is not returnable.
View return policy
Description
- Objectives with long working distance for incident light provide optimal image contrast for reflective, non-covered specimen
- Together with a fully corrected tube lens, an intermediate image without colored fringes is created
- 0.8 N.A.
Specifications
LM Plan Achromatic Objective | |
BA310MET Series | |
100X | |
2 mm |