missing translation for 'onlineSavingsMsg'
Learn More

Lithium for AAS 10g/L HNO3 matrix, Fisher Chemical™

Artikelnummer. 12913335
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
Packaging:
Bottle
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Spezifikation

Concentration 10g/L ±0.02g/L
CAS 7697-37-2
Packaging Bottle
Quantity 100 mL
Type Single-element Concentrated Standard Solution
Molecular Formula HNO3
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES O[N+]([O-])=O
IUPAC Name nitric acid
Molecular Weight (g/mol) 63.01
PubChem CID 944
ChEBI CHEBI:48107
Solvent or Matrix 2% HNO3
Physical Form Liquid
Chemical Name or Material Lithium
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • Single-element Standard Solution for AAS. Lithium (Li) 10000mg/l in HNO3 2%
Signalwort
  • Warning
Gefahrenkategorie
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Sicherheitshinweis
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'