Learn More
Sampling Systems™ Liquid Master
Product Code. 15869331 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
150 mL
Unit Size:
Each
Product Code. 15869331
Brand: Sampling Systems™ 5035S600
Only null left
Add to basket
Add to basket
Available in a range of lengths and diameters to give a range of sample volumes. Sampling Systems™ Liquid Master is ideal for taking samples from low and medium viscosity liquids.
  • Product Contact Material Available in: 316L Stainless Steel, Food Grade PTFE/FEP, Polypropylene
  • Easily stripped down for cleaning
  • Single Point and Cross Sectional Sampling
Specifications
Product Type Liquid Sampler
Material Stainless Steel (316L)
Capacity (Metric) 150 mL
For Use With (Application) Sampling Liquids
Diameter (Metric) Outer 25 mm
Length (Metric) 600 mm