Learn More
Kartell™ Lining for Glass Tapers
Specially designed to prevent machined glass joints from binding
Brand:  Kartell™ 0104200
Product Code. 15835497
  • € 34.80 / Pack of 10
Estimated Shipment: 01-05-2025
to see stock.
Explore more special offers
Description
  • Conforms to DIN 12242
  • Suitable for foodstuff
Compliance
DIN 12242
Specifications
Specifications
PTFE
10/pk
31 mm
34/35
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Kartell™ Lining for Glass Tapers > Cone size: 34/ 35