Learn More
Limonin, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
Limonin is a triterpenoid enriched in citrus fruits, which has antivirus and antitumor ability.
Specifications
Specifications
| Chemical Name or Material | Limonin |
| CAS | 1180-71-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C26H30O8 |
| Quantity | 100 mg |
| Synonym | Limonoic acid 3,19:16,17 dilactone |
| Solubility Information | DMSO : 41.67 mg/mL (88.56 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C1[C@@H]2[C@@]3(O2)[C@](CC[C@@]([C@@]4([C@@](CC(OC4)=O)([H])OC5(C)C)[C@@]5([H])CC6=O)([H])[C@]36C)(C)[C@H](C7=COC=C7)O1 |
| Molecular Weight (g/mol) | 470.51 |
| Show More |
Research purposes only