Learn More
Gilac™ Lightweight Nesting Pallet
Click to view available options
Dimensions (L x W x H):
600 x 800 x 130 mm
800 x 1200 x 155 mm
1200 x 1000 x 160 mm
Unit Size:
Each
Description
- Lightweight nesting plastic pallet.
- Nesting and therefore space-saving when stored empty.
- Storage prohibited on racks: the pads of the pallets are incompatible with the rails of the racks.
Specifications
Specifications
| Product Type | Aero Tote Pallet |
| Color | Black |
| Dimensions (L x W x H) | 800 x 1200 x 155 mm |
| Capacity (English) | 3306.93 lb. |
| Capacity (Metric) | 1500 kg |
| Height (English) | 6.1 in. |
| Height (Metric) | 155 mm |
| Length (English) | 31.5 in. |
| Length (Metric) | 800 mm |
| Width (English) | 47.2 in. |
| Show More |