missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Atto565 Antibody Labeling Kit
Applications: Flow Cytometry
Brand: Novus Biologicals™ 351-0030
This item is not returnable.
View return policy
Description
Atto565 is one of a new generation of fluorescent labels. It has a strong absorption at 563nm, high fluorescence at 592nm (extinction coefficient 1.2 x10&superscript_begin;5&superscript_end; cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and quantum yield.Order Info
Quantity

Specifications
Store at -20°C. Avoid freeze-thaw cycles. | |
30 μg | |
Atto 565 | |
Rapid Atto565 Antibody Labeling Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Lightning-Link (R) Rapid Atto565 Antibody Labeling Kit > 3 x 20μg Reactions
Spot an opportunity for improvement?Share a Content Correction