missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Light Module Cold White 6,500 K + Warm White 2,700 K

Product Code. p-7220936 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP110eco
HPP260eco
HPP410eco
HPP750eco
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17109804

Brand: Memmert™ HPP260ecoT8

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

LED light strips on the side walls of the interior

  • 6 resp. 8 alternating cold white light strips and 5 resp
  • 7 warm white light strips
  • On the side walls of the interior
  • Programme-controlled dimming from 0 to 100 % (in 1 % steps)
  • Ramp programming in combination with temperature and humidity
  • Only withTwinDISPLAY; not in combination with entry ports F6, F7, D8

Specifications

Color White
For Use With (Equipment) HPP260eco
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.