missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Light Module Cold White 6,500 K

Product Code. 17175324 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP260eco
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17175324

Brand: Memmert™ IPP260ECOT7

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Light strips arranged on the side walls of the interior

  • 10 strips for model 110
  • 14 strips for model 260/410/750
  • Programme-controlled dimming from 0 to 100 %(in 1 % steps)
  • Ramp programming in combination with temperature
  • Only with TwinDISPLAY; not in combination with entry ports F6, F7, D8

Specifications

Color White
For Use With (Equipment) IPP260eco
For Use With (Application) Only with TwinDISPLAY
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.