Learn More
Memmert™ Light Module Cold White 6,500 K
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP260eco
Unit Size:
Each
Description
- 10 strips for model 110
- 14 strips for model 260/410/750
- Programme-controlled dimming from 0 to 100 %(in 1 % steps)
- Ramp programming in combination with temperature
- Only with TwinDISPLAY; not in combination with entry ports F6, F7, D8
Specifications
Specifications
| Color | White |
| For Use With (Equipment) | HPP110eco |
| For Use With (Application) | Only with TwinDISPLAY |