Learn More
Invitrogen™ Novex™ IL-6 Human Ultrasensitive Magnetic Bead Kit
Designed for quantification of low levels of human IL-6 in serum, plasma and tissue culture supernatant samples. Invitrogen™ Novex™ IL-6 Human Ultrasensitive Magnetic Bead Kit is an ideal solution for assessing multiple protein biomarkers in a single sample.
Brand: Invitrogen™ LHC0063M
Includes: Antibody coated capture beads, standard, detector antibody and lot-specific technical data sheet
Description
This immunoassay uses Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins. This assay is subject to the same stringent and robust development and manufacturing as are the extensively validated Platinum and Instant ELISA kits. Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma.