Learn More
Fisherbrand™ Lids for Desiccator with nz 24/29
For use with desiccators with nz 24/29
€ 86.80 - € 255.28
Product Code | Brand | Diameter (Metric) Outer | Height (Metric) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Height (Metric) | Description | Price | Quantity & Availability | ||||
15574635
|
Fisherbrand™
632415280252 |
320 mm | 111 mm | Type: 250 mm |
€ 193.44
Each |
In Stock
Log in to see stock. |
||||
15574625
|
Fisherbrand™
632415280152 |
215 mm | 89 mm | Type: 150 mm |
€ 102.00
Each |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||
15524645
|
Fisherbrand™
632415280302 |
380 mm | 136 mm | Type: 300 mm |
€ 255.28
Each |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||
15524635
|
Fisherbrand™
632415280202 |
270 mm | 97 mm | Type: 200 mm |
€ 164.00
Each |
Estimated Shipment: 30-06-2025
Log in to see stock. ![]() |
||||
15524625
|
Fisherbrand™
632415280102 |
153 mm | 64 mm | Type: 100 mm |
€ 86.80
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||
Description
- For use with desiccators with nz 24/29
Certifications
ISO 13 130, DIN 12 491
Specifications
Lid | |
Desiccator |
Product Title
Fisherbrand™ Lids for Desiccator with nz 24/29