Learn More
Lidocaine-d10, Honeywell Fluka™ Product Code.: 15920252
Lidocaine-d10, Honeywell Fluka™
Product Code. 15920252 Shop All Honeywell Fluka Products
10 mg
Click to view available options
Quantity:
10 mg
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15920252
Brand: Honeywell Fluka R151610MG
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
CAS 851528-09-1
Packaging Glass Bottle
Quantity 10 mg
For Use With (Application) Anesthetics
Physical Form Solid
Grade Analytical Standard
Product Content Correction
Título del producto
Lidocaine-d10, Honeywell Fluka™ >
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado