Learn More
Liebherr™ LGv 5010 MediLine Freezer
Protects valuable contents with a variety of alarm, memory and safety features. Liebherr™ LGv 5010 MediLine Freezer features the Comfort electronic controller, advanced cooling technology, multiple organizing drawers/bins, and an easy-clean molded plastic interior.
Brand:  Liebherr™ LGV 5010
Product Code. 15786918
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Integrated Alarms:
  • Audio and visual temperature, door opening and power failure alarms, as well internal system alarms (e.g. probe)
  • Alarm Test Function: Check whether an internally or externally connected alarm system is functioning correctly without impacting refrigeration performance
  • Comfort Controller with Digital Display: Internal memory stores details of last three alarms and high/low temperatures
  • Serial Bus Interface: Electronic control system uses an RS 485 interface to connect up to 20 appliances to external monitoring software
  • Temperature Stability EN 60068-3: Measures the temperature at nine points in the interior according to procedure EN 60068-3, using state-of-the-art facilities
  • Forced Air Cooling: Highly efficient fans quickly achieves and maintains an even temperature throughout the interior
  • Efficient Refrigeration System: Reduces energy consumption and operating costs with premium-quality components
  • Environmentally Friendly Refrigerant: Minimal greenhouse effect and no ozone depletion potential with outstanding energy efficiency