missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Leupeptin, 90%, synthetic Product Code.: 10697934
Add to basket

Thermo Scientific Chemicals Leupeptin, 90%, synthetic

Product Code. 10697934
100 mg
Click to view available options
Quantity:
5 mg
25 mg
100 mg
This item is not returnable. View return policy

Product Code. 10697934

Brand: Thermo Scientific Chemicals 328351000

Add to basket
Add to basket
Request Bulk or Custom Format
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103476-89-7
Molecular Formula 0·5 H2O4S

Specifications

Color White
Product Type Leupeptin
Formula Weight 475.6
Solubility Solubility in water: soluble in water
Form Powder
Quantity 100 mg
Percent Purity 90%
Chemical Name or Material Leupeptin
compliance-icons
Product Identifier
  • Leupeptin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Leupeptin, 90%, synthetic >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.