missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) cyclohexanebutyrate, 94%, Thermo Scientific™

Product Code. p-7023577
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11473507

Brand: Thermo Scientific Alfa Aesar A17280.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62637-99-4
Molecular Formula C20H34O4Pb
Molecular Weight (g/mol) 545.688
MDL Number MFCD00036403
InChI Key CCJZHCZMNGUOCA-UHFFFAOYSA-L
Synonym lead cyclohexanebutyrate, lead bis 4-cyclohexylbutyrate, cyclohexanebutanoic acid, lead 2+ salt, cyclohexanebutanoic acid, lead 2+ salt 2:1, lead 2+ bis 4-cyclohexylbutanoate, acmc-20akns, lead ii cyclohexanebutyrate, 4-cyclohexylbutanoate; lead 2+, biscyclohexanebutyric acid lead ii salt, $l^ 2-lead 2+ ion bis 4-cyclohexylbutanoate
PubChem CID 112896
IUPAC Name 4-cyclohexylbutanoate;lead(2+)
SMILES C1CCC(CC1)CCCC(=O)[O-].C1CCC(CC1)CCCC(=O)[O-].[Pb+2]

Specifications

Quantity 5 g
UN Number UN2291
Formula Weight 545.68
Percent Purity 94%
Chemical Name or Material Lead(II) cyclohexanebutyrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) cyclohexanebutyrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.