Learn More
Vitlab™ LDPE Dropper Bottles
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
Unit Size:
Pack of 12
Pack of 24
Description
- Narrow-mouth bottles
- Made of LDPE
- Transparent
- With dropper insert
- Screw cap made from LDPE
- Extra long, fine dropping tip
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Color | Transparent |
| Material | LDPE |
| No. per Pack | 12Pack |
| Quantity | 12Pack |
| Mouth | Narrow |
| Height (Metric) | 183 mm |
| Product Type | Dropping Bottle |
| Diameter (Metric) | 58 mm |