missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-AKT [pSer473] Antibody Kit
LanthaScreen™ Tb-labeled phospho-specific antibodies allow for the detection of intracellular post-translational modifications of specific target proteins.
Brand: Thermo Scientific™ PV5268
This item is not returnable.
View return policy
Description
- LanthaScreen™ Tb-labeled phospho-specific antibodies allow for the detection of intracellular post-translational modifications of specific target proteins
- To facilitate in-house kinase assay configuration and screening, we have developed a selection of Tb-labeled phospho-specific antibodies
- These antibodies may be accessed in small scale and screening quantities
Specifications
Monoclonal | |
Tb (Terbium)/Lanthanide Chelates | |
Purified | |
Terbium labeled anti-AKT [pSer473] antibody in HEPES buffered saline (137 mM NaCl, 2.7 mM KCl, 10 mM HEPES pH 7.5). Storage is at -20°C | |
IgG | |
Rb | |
Human | |
Hu |
0.5 mg/mL | |
Rabbit | |
1 mg | |
Liquid | |
Microplate Reader | |
0.5 mg/mL | |
Dry Ice | |
LanthaScreen™ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction