Learn More
Brady™ Laminated Polyester Warning Safety Signs
Product Code. 13586790
Click to view available options
Width (Metric):
50 mm
100 mm
200 mm
315 mm
Displayed Text:
Danger Zone Warning Pictogram - !
Warning about skid risk
Warning of cold
Warning of industrial trucks
Warning of tripping hazard
Unit Size:
Each
Product Code. 13586790
Brand: Brady™ 250270
Only null left
Specifications
Certifications/Compliance ISO 7010, RoHS compliant using EU Directive 2002/95/EC, ROHS Environmental Compliance
Alternate Part Number PIC 323-TRI 100-B7541
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Description Laminated Polyester
For Use With (Application) Pipe markers and safety signs
Height (Metric) 100 mm
Thickness 0.024 mm
Width (Metric) 100 mm
Temperature Range (Metric) -40°C to 110°C
Shape Triangular
Displayed Text Warning of cold
Product Type Warning Safety Sign
Color Black on Yellow