missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Laminated Polyester Warning Safety Signs
Click to view available options
Width (Metric):
50 mm
100 mm
200 mm
315 mm
Displayed Text:
Danger Zone Warning Pictogram - !
Warning about skid risk
Warning of cold
Warning of industrial trucks
Warning of tripping hazard
Unit Size:
Each
Specifications
Specifications
| Certifications/Compliance | ISO 7010, RoHS compliant using EU Directive 2002/95/EC, ROHS Environmental Compliance |
| Alternate Part Number | PIC 306-TRI 050-B7541 |
| Material | Polyester |
| Quantity | 1 Ea. |
| Adhesive Type | Permanent Acrylic |
| Description | Laminated Polyester |
| For Use With (Application) | Pipe markers and safety signs |
| Height (Metric) | 50 mm |
| Thickness | 0.024 mm |
| Width (Metric) | 50 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction