accessibility menu, dialog, popup
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Facility Safety and Maintenance
Signs and Tags
Facilities Maintenance Signs
Brady™ Laminated Polyester Prohibition Signs: Do Not Walk Under Lifted Load
Print
Brady™ Laminated Polyester Prohibition Signs: Do Not Walk Under Lifted Load
€ 3.30 - € 4.20
Specifications
Shape
Round
View More Specs
Products
2
Documents
Product Certifications
Products
2
Product Code
Brand
Diameter (Metric)
Price
Quantity & Availability
Product Code
Brand
Diameter (Metric)
Price
Quantity & Availability
10401264
View Documents
missing translation for 'viewProductCertificates'
Brady™
250031
100 mm
€ 3.30
Each
In Stock
Log in
to see stock.
Add to basket
10253784
View Documents
missing translation for 'viewProductCertificates'
Brady™
250032
200 mm
€ 4.20
Each
Estimated Shipment: 04-12-2024
Log in
to see stock.
Specifications
Shape
Round
Documents
Product Certifications
Back to Top