Learn More
Brady™ Laminated Polyester Mandatory Signs
Product Code. 10451264
Click to view available options
Diameter (Metric):
100 mm
200 mm
310 mm
Displayed Text:
Direction Note
Use Breathing Equipment
Use Breathing Protection
Use Eye and Ear Protection
Use Face Shield
Use Foot Protection
Use Full Protective
Use Head and Hearing Protection
Use Overshoes
Use Protective Clothing
Use Protective Hood
Use Safety Coat
Use and Respiratory Protection - Head
Use eye protection
Use head protection safety
Use hearing protection
Use respiratory protection
Wear Protective Footwear
wear Conductive Footwear
Unit Size:
Each
Product Code. 10451264
Brand: Brady™ 250138
Only null left
Add to basket
Add to basket
Mandatory signs to assist with personal safety
Sign, mandatory - Hair Cap Required
  • Complies to NF X 08-003-1 and 08-003-3 N X and EU Directive 92/58/EEC
  • Resistant to chemicals; water; UV light; scratch/abrasion and temperatures up to 120°C
  • Diameter 100mm
  • Blue or white
Specifications
Alternate Part Number PIC 269-DIA 200-B7541 (BRANDED)
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Description Laminated Polyester
Diameter (Metric) 200 mm
Max. Temperature (Metric) 110°C
Thickness 0.024 mm
Shape Round
Displayed Text Use Protective Hood
Product Type Mandatory Sign
Color White on Blue