Learn More
Brady™ Laminated Polyester GHS Symbol Signs
Product Code. p-8001106
Click to view available options
Width (Metric):
40 mm
50 mm
70 mm
100 mm
Displayed Text:
Acute Toxicity
Compressed Gas
Corrosive
Explosive
Flammable
Hazardous to Aquatic Environment
Health Hazard
Oxidizing
Respiratory Hazard
Style:
Card Mounted
Continuous Roll, 250 Symbols (not labels)/Roll
Individual Markers on a Roll
Quantity:
1 Ea.
100/Pk.
20/Pk.
250/Pk.
6/Pk.
Unit Size:
1 roll
Pack of 20
Pack of 250
Pack of 4
Pack of 6
Product Code. 15414871
Brand: Brady™ 811720
Only null left
Add to basket
Edge
Add to basket
Help ensure safety at work and maximize productivity with these GHS warning labels. Brady™ Laminated Polyester GHS Symbol Signs are available in a variety of sizes, with and without text. Standardized images reduce the risk of misunderstandings.
  • Self-adhesive diamond shape labels
  • Pre-printed with GHS hazard symbols
  • Laminated polyester, gloss finish black and red on white
Specifications
Certifications/Compliance ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Alternate Part Number PIC 1809-40*40-B7541-CRD
Finish Gloss
Material Polyester
Quantity 20/Pk.
Style Card Mounted
Adhesive Type Permanent Acrylic, Pressure-sensitive
Description Laminated Polyester
For Use With (Application) Pipe markers and safety signs
Height (Metric) 40 mm
Thickness 0.024 mm
Width (Metric) 40 mm
Temperature Range (Metric) -40°C to 110°C
Shape Diamond
Displayed Text Acute Toxicity
Product Type GHS Symbol Sign
Color Black on White