accessibility menu, dialog, popup
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Facility Safety and Maintenance
Signs and Tags
Facilities Maintenance Signs
Brady™ Laminated Polyester First Aid Signs: Stretcher Pictogram
Print
Brady™ Laminated Polyester First Aid Signs: Stretcher Pictogram
€ 3.30 - € 4.90
Specifications
Finish
Gloss
View More Specs
Products
2
Documents
Product Certifications
Products
2
Product Code
Brand
Dimensions (L x W)
Price
Quantity & Availability
Product Code
Brand
Dimensions (L x W)
Price
Quantity & Availability
10586654
View Documents
missing translation for 'viewProductCertificates'
Brady™
250415
100W x 100 mmH
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10356154
View Documents
missing translation for 'viewProductCertificates'
Brady™
250417
200W x 200 mmH
€ 4.90
Each
Estimated Shipment: 11-12-2024
Log in
to see stock.
Specifications
Finish
Gloss
Documents
Product Certifications
Back to Top