missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Laminated Polyester Biohazard Warning Signs

Product Code. 10795535
Click to view available options
Description:
Oblong
Triangular, 100W x 87 mmH
Triangular, 200W x 173 mmH
Triangular, 50W x 43 mmH
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10795535

Brand: Brady™ 250238

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Alternate Part Number PIC 312-TRI 200-B7541 (BRANDED)
Finish Gloss
Material Laminated Polyester
Adhesive Type Permanent Acrylic
Description Triangular, 200W x 173 mmH
Height (Metric) 173 mm
Width (Metric) 200 mm
Shape Triangular
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.