Learn More
Brady™ Laminated Polyester Anti-skid Tape-Die-Cut Cleats
Laminated Polyester Anti-skid Tape-Die-Cut Cleats help to prevent slipping and falling on wet or slippery floors.
Brand:  Brady™ 078162
 View more versions of this product
Product Code. 11796925
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Anti-Skid (anti-slip) tapes help to prevent slipping and falling on wet or slippery floors.
  • The rugged polyester tape is adhesive backed and coated with thousands of particles of durable aluminium oxide grit.
  • Anti-Skid tape bonds permanently to clean, dry floors, withstands grease and oil and will work indoors or outdoors.
  • Anti-Skid tape is available in a variety of colors and sizes to suit all of your floor marking needs
Specifications
Specifications
ANSI A1264.2-2006, NSFI Certified for High Traction, OSHA 1910.144, OSHA 1910.22
150 x 600 mm
5.9 in.
24 Pk.
-40°C to +70°C
24 in.
Permanent Acrylic
Polyester
Glow-in-the-Dark Die-Cut Cleats - Black
Traction on steps, slick floors, and egress pathway marking
150 mm
-40°F to +158°F
0.162 mm
600 mm
Photoluminescent, Black
Anti-skid Tape
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers