Learn More
Thermo Scientific™ Labtip Pipette Tips
GREENER_CHOICE
Product Code. p-6445572
Click to view available options
Volume (Metric):
200 μL
1000 μL
Format:
Bulk
Rack
Packaging:
10 x 96/Tray
10,000 tips/bag
1000 tips/bag
25,000 tips/bag
Unit Size:
Pack of 1000
Pack of 10000
Pack of 25000
Pack of 3840
Product Code. 12326582
Brand: Thermo Scientific™ 94300210
Only null left
Add to basket
Add to basket
Basic pipette tips for standard liquid transfer needs.
Thermo Scientific™ Labtip standard pipette tips are made from 100% virgin polypropylene. These tips are available in 10, 200, 300 and 1000 μL sizes. Available in clear (10 μL and 300 μL), yellow (200 μL), blue (1000 μL), these tips for general lab use across countless liquid transfer applications.
Labtip pipette tips provide:
  • The most common sizes used in a lab: 10, 200, 300 and 1000 μL
  • Non-Filter essential tips for general laboratory usage
  • Non-sterile packaging in both Racked and Bulk options
Packaging Options:
  • Racked, nonsterile - 96 tips/ tray, 10 trays/pack
  • Bulk - 1,000 tips/ bag
  • Bulk - 25,000 tips/bag
TRUSTED_SUSTAINABILITY
Specifications
Description Labtip 1000, Non-filtered, Non-sterile, Bulk
Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Blue
Autoclavable Not Autoclavable
Model Labtip 1000
Clean Claims DNA, Dnase, RNase free
Features blue tip
Format Bulk
Tip Style Traditional
Packaging 10,000 tips/bag
Quantity 10000
Disposable Single-use
For Use With (Equipment) Universal Fit
Product Line LabTip
Material Virgin Polypropylene