Learn More
Thermo Scientific™ Labtainer™ 2D BioProcess Container (BPC), three ports, MPCs, quick-connect
Thermo Scientific Labtainer 2D BioProcess Containers (BPCs) are composed of Aegis5-14 single-web, multi-layer film.
€ 588.64 - € 1433.20
Specifications
Autoclavable | Not Autoclavable |
---|---|
Certifications/Compliance | ISO 7 |
Color | Transparent |
Disposable | Single-use |
For Use With (Application) | Bioprocessing |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17850597
|
Thermo Scientific™
SH31005.01 |
5 L |
€ 713.76
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17870597
|
Thermo Scientific™
SH31005.02 |
10 L |
€ 753.57
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17810607
|
Thermo Scientific™
SH31005.03 |
20 L |
€ 807.60
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17890597
|
Thermo Scientific™
SH31005.04 |
50 L |
€ 1433.20
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17800607
|
Thermo Scientific™
SH31005.05 |
2 L |
€ 588.64
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Features of the Labtainer 2D BPC include:
• Helps eliminate post-use cleaning steps required with re-useable containers and reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions
Applications of the Labtainer 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Prepare and store buffer, culture media, and process liquid
• Collect chromatography feed and fractions
• Harvest, storage, and transport bulk drug product and bulk drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands
Specifications
Not Autoclavable | |
Transparent | |
Bioprocessing | |
C-Flex | |
3 ports |
ISO 7 | |
Single-use | |
Sterile | |
Aegis 5-14 |