Learn More
Nickel Electro™ Clifton™ Nickel Crucible Lids
Shop All Nickel-Electro Products
Click to view available options
Diameter (Metric) Outer:
20 mm
35 mm
40 mm
45 mm
50 mm
60 mm
Unit Size:
Each
Description
- Provides a cover to crucibles
- Prevents chance of hot material spattering out of open crucible during heating process
- Lid can be easily removed with either stainless steel or nickel laboratory tongs
- Forms a tightly adhering oxide file between 400°C and 600°C in oxidizing atmospheres
- Nickel effective for use in fusions where zirconium or other metals cannot be used
Specifications
Specifications
| Material | Nickel |
| Max. Temperature (Metric) | 350°C |
| Quantity | Each |
| Diameter (Metric) Outer | 60 mm |
| Product Type | Laboratory Crucible Lid |