missing translation for 'onlineSavingsMsg'
Learn More

Laboratory Fortifying Stock Solution 1, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep LFS-1-100N


 View more versions of this product

Product Code. 15226106

  • € 397.00 / 125mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
This item is not returnable. View return policy

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
P50μg/mL
Al25μg/mL
As25μg/mL
B25μg/mL
Ba25μg/mL
Cr25μg/mL
Cu25μg/mL
Fe25μg/mL
Li25μg/mL
Mn25μg/mL
Ni25μg/mL
Pb25μg/mL
Sb25μg/mL
Se25μg/mL
SiO&subscript_2;25μg/mL
Sr25μg/mL
Tl25μg/mL
Zn25μg/mL
Cd10μg/mL
Co10μg/mL
Mo10μg/mL
Sn10μg/mL
V10μg/mL
Be5μg/mL
Ag2.5μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

LFS Solution 1 without Mercury
125 mL
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
5% HNO3/tr Tart. Acid/tr HF
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
50μg/mL: P; 25μg/mL: Al,As,B
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • LFS Solution 1

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : nitric acid, hydrofluoric acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product intended for laboratory use only