Learn More
3M™ Zeta Plus™ Laboratory Capsule ZB Series Media 25 cm2
Zeta Plus™ BC Series Filter Capsules with ZB Series Media are ideal for bio-processing, biological, and small molecule pharmaceutical manufacturing applications where an optimized balance between throughput and particle removal efficiency is critical.
Brand: 3M™ Zeta Plus™ 7100102711
This item is not returnable.
View return policy
Description
- Practical and easy to use tool to evaluate the Zeta Plus Depth Filter range
- Large range of dual layer media combinations are offered resulting in highly gradient porosity structures that can be optimized for throughput and removal efficiency in applications from cell culture harvest to fine clarification
- BC25 Laboratory capsules contain same media as in production scale systems and are ideal for selecting the optimum media grade and media grade combination using low challenge fluid volumes
- The capsules are ideal for selecting media grades using low fluid volumes
- ZB series have the highest anion capacity from all Zeta Plus range, allowing for the removal of negatively charged contaminants
Specifications
Autoclavable | |
Non-sterile | |
50 to 200 LMH | |
2.4 bar | |
40°C |
Cellulose, Silica, Polymer Resin | |
25 cm2 | |
Luer-Lock | |
4/Pk. |