Learn More
Bochem™ Laboral Retort Clamp, Flexible Shaft
Coated with soft silicone, the three fingers of this retort clamp are well suited for holding delicate objects. Laboral Retort Clamp, Flexible Shaft has a bendable support shaft for positioning ease.
Brand:  Bochem™ B08309658
Product Code. 10320301
  • € 208.00 / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Description
  • Laboral aluminum alloy construction.
  • Silicone-coated fingers.
  • Adjustable span width.
  • Nickel-plated, flexible shaft design.
Specifications
Specifications
Laboral, Silicone
225 mm