Learn More
Buerkle™ LaboPlast™ Spoon Spatulas
BIOPHARMAGREENER_CHOICE
Use as a sampling spoon or to measure or transfer small amounts. Buerkle™ LaboPlast™ Spoon Spatulas can also be used as a scraper or for mixing and stirring.
Brand:  Buerkle™ 5378-0031
31.26 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Product Code. 15674104
  • € 32.90 / Pack of 100
In Stock
to see stock.
Explore more special offers
Description
  • Polystyrene, white
  • Specifications
    Specifications
    Polystyrene
    Single-use
    17 mm spoon blade
    0.5 mL
    Non-sterile
    Videos
    • Buerkle LaboPlast Spoon Spatulas
      Buerkle LaboPlast Spoon Spatulas
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Buerkle LaboPlast Spoon Spatulas
      SDS
      Documents Certificates
      Certificates
      A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
      5 results found
      Lot Number Certificate Type Date Catalog Number
      Lot Number250358 Certificate TypeCertificate of Compliance Date04/02/2025 Catalog Number5378-0031
      Lot Number243843 Certificate TypeCertificate of Compliance Date26/09/2024 Catalog Number5378-0031
      Lot Number242041 Certificate TypeCertificate of Compliance Date10/06/2024 Catalog Number5378-0031
      Lot Number240879 Certificate TypeCertificate of Compliance Date12/03/2024 Catalog Number5378-0031
      Lot Number233215 Certificate TypeCertificate of Compliance Date03/08/2023 Catalog Number5378-0031
      Special Offers
      Special Offers
      Product Content Correction
      Product Title
      Buerkle™ LaboPlast™ Spoon Spatulas > 0.5 mL