Learn More
Buerkle™ LaboPlast™ Bio Disposable Scoop
Shop All Bürkle Products
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
150 mL
Unit Size:
Pack of 10
Description
- Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled
- The scoop stands horizontally as a result of the special handle design and the slightly angled front edge and can therefore be put down without the sample material trickling out.
- Product and packaging made of renewable raw materials
- Bio-PE, white
- Individually wrapped
- Production and packaging according to Cleanroom Class 7
- With EU foodstuffs and FDA approval
Specifications
Specifications
| Quantity | 10 |
| Sterility | Non-sterile |
| Capacity (Metric) | 150 mL |
| Color | White |
| Certifications/Compliance | FDA |
| Disposable | Single-use |
| For Use With (Application) | Foodstuffs |
| Material | Polyethylene |
| Length (Metric) | 216 mm |