Learn More
Hirschmann™ labopette™ 8-Channel Pipette
Click to view available options
Volume (Metric):
0.5 to 10 μL
10 to 100 μL
30 to 300 μL
Accuracy:
1.6%
2.4%
Compatible Tips:
Clear, 0.2-10 μL
Clear, 1.0-250 μL
Clear, 5.0-300 μL
Increments:
0.01 μL
0.1 μL
0.2 μL
Unit Size:
Each
Description
- Variable volume setting
- Multi channel
- With tip ejector and recalibration possibility
- Autoclavable
Specifications
Specifications
| Accuracy | 1.6% |
| Certifications/Compliance | ISO 8655 |
| Compatible Tips | Clear, 1.0-250 μL |
| Description | Tip ejector |
| Autoclavable | Autoclavable |
| Number of Channels | 8 |
| Product Line | Labopette |
| Type | Multichannel Pipette with Variable Volume Setting |
| Volume (Metric) | 10 to 100 μL |
| Includes | Calibration device, individual number and weighing protocol |
| Show More |