missing translation for 'onlineSavingsMsg'
Learn More

MHC II (HLA-DR) Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ MHC II (HLA-DR) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-133-P1

 View more versions of this product

Product Code. 12639616

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

HLA-DR, like other MHC class II molecules, is a transmembrane glycoprotein composed of an α chain (36 kDa) and a β chain (27 kDa). It is expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides and present them to T-helper cells. They therefore have a critical role in the initiation of the immune response.

Host Species: Mouse

Clone: LN3

Isotype: IgG2b

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Activated human peripheral blood mononuclear cells

Molecular Weight: 36kDa (α-chain) and 27kDa (β-chain)

Positive Control: Tonsil/lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

MHC II (HLA-DR) Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Hematopoietic
Human
IgG2b
Flow Cytometry, Immunohistochemistry (Paraffin)
LN3
Unconjugated
Activated human peripheral blood mononuclear cells
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.