missing translation for 'onlineSavingsMsg'
Learn More

β-Catenin Rabbit Polyclonal Antibody, Epredia™

Stain formalin-fixed, paraffin-embedded tissues with Epredia™ Catenin, β, Rabbit Polyclonal Antibody.

Brand:  Epredia RB-9035-R7

 View more versions of this product

Product Code. 12693377

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

The catenins (α, Β and γ) are ubiquitously expressed cytoplasmic proteins associated with E-cadherin at cellular junctions. Β-catenin also binds to N-cadherin and co-immunoprecipitates with APC. Cadherin/catenin complexes are linked to the cytoskeleton via a direct association between α-actinin and α-catenin. Increased tyrosine phosphorylation can disrupt catenin-cadherin complexes, influencing cellular adhesion.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of the β-catenin protein

Molecular Weight: 92kDa

Positive Control: MCF-7 cells, Breast CA

Cellular Localization: Cytoplasmic and cell membrane

This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation
Specifications

Specifications

Catenin, beta
Polyclonal
Unconjugated
A synthetic peptide from the C-terminus of the beta-catenin protein
IVD
Primary
Prediluted
Immunoprecipitation, Western Blot
Polyclonal
Rabbit
7 mL
Transcription Regulation
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.