missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Threonine methyl ester hydrochloride, 98%
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Description
L-Threonine methyl ester hydrochloride is used as a feed and food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Threonine methyl ester hydrochloride is used as a feed and food additive.
Solubility
Soluble in water.
Notes
Moisture Sensitive and Hygroscopic. Keep container tightly closed. Store away from water/moisture.
L-Threonine methyl ester hydrochloride is used as a feed and food additive.
Solubility
Soluble in water.
Notes
Moisture Sensitive and Hygroscopic. Keep container tightly closed. Store away from water/moisture.
Specifications
Specifications
| Beilstein | 3562717 |
| Solubility Information | Soluble in water. |
| Quantity | 5 g |
| Formula Weight | 169.61 |
| Sensitivity | Hygroscopic; Moisture sensitive |
| Percent Purity | 98% |
| Chemical Name or Material | L-Threonine methyl ester hydrochloride |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction