Learn More
L-Hyoscyamine, MedChemExpress

MedChemExpress L-Hyoscyamine (Daturine), a natural plant tropane alkaloid, is a potent and competitive muscarinic receptor (MR) antagonist. L-Hyoscyamine is a levo-isomer to Atropine (HY-B1205).
POA - € 112.50
Specifications
Chemical Name or Material | L-Hyoscyamine |
---|---|
CAS | 101-31-5 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C17H23NO3 |
Description
L-Hyoscyamine (Daturine), a natural plant tropane alkaloid, is a potent and competitive muscarinic receptor (MR) antagonist. L-Hyoscyamine is a levo-isomer to Atropine (HY-B1205).Specifications
L-Hyoscyamine | |
White | |
C17H23NO3 | |
DMSO : 100 mg/mL (345.58 mM; Need ultrasonic) | |
289.37 | |
98.2% | |
Solid |
101-31-5 | |
Research | |
Daturine | |
CN1[C@@H]2C[C@@H](OC([C@@H](C3=CC=CC=C3)CO)=O)C[C@H]1CC2 | |
289.37 | |
Research | |
Neuroscience-Neurodegeneration |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.