Learn More
Thermo Scientific Chemicals L-(+)-Asparagine monohydrate, 98+%
Description
L-Asparagine is a naturally occurring proteinogenic amino acid, used in biomanufacturing cell culture systems for the production of therapeutic recombinant proteins and monoclonal antibodies. It is used in the biosynthesis of proteins. It is also required for development and function of the brain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-Asparagine is a naturally occurring proteinogenic amino acid, used in biomanufacturing cell culture systems for the production of therapeutic recombinant proteins and monoclonal antibodies. It is used in the biosynthesis of proteins. It is also required for development and function of the brain.
Solubility
Soluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∼245°C (decomposition) |
| Beilstein | 5767869 |
| Merck Index | 14,837 |
| Solubility Information | Soluble in water. |
| Density | 1.543 |
| Quantity | 500 g |
| Formula Weight | 150.14 (132.12 Anhydrous) |
| Optical Rotation | +31° (c=5 in 5N HCl) |
| Percent Purity | ≥98% |
| Chemical Name or Material | L-(+)-Asparagine monohydrate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.