missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid, Certified AR for Analysis, Fisher Chemical

Product Code. p-7072846 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10713791

Brand: Fisher Chemical A/8882/48

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

C6H8O6, CAS Number-50-81-7, 500g, White, 176.13g/mol, 190 deg.C, 64328, Plastic powder jar, 2.1 to 2.6, Solid | CAS: 50-81-7 | C6H8O6

C6H8O6, CAS Number-50-81-7, 500g, White, 176.13g/mol, 190 deg.C, 64328, Plastic powder jar, 2.1 to 2.6, Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
MDL Number 64328

Specifications

Melting Point 190°C
Color White
pH 2.1 to 2.6
Packaging Plastic powder jar
Quantity 100 g
Formula Weight 176.13g/mol
Physical Form Solid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.