missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KV1.8 (KCNA10) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541032
This item is not returnable.
View return policy
Description
Peptide sequence: PANVPIDIFA DEISFYELGS EAMDQFREDE GFIKDPETLL PTNDIHRQFW Sequence homology: Cow: 100%; Dog: 92%; Guinea Pig: 92%; Human: 100%; Mouse: 100%; Rat: 100%
KV1.8 (KCNA10) Polyclonal antibody specifically detects KV1.8 (KCNA10) in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
KV1.8 (KCNA10) | |
Polyclonal | |
Unconjugated | |
KCNA10 | |
cyclic GMP gated potassium channel; Gm1962; Kcn1; KCNA10; Kcna8; Kv1.8; potassium channel, voltage gated shaker related subfamily A, member 10; potassium voltage gated channel, shaker related subfamily, member 8; potassium voltage-gated channel subfamily A member 10; potassium voltage-gated channel, shaker-related subfamily, member 10; potassium voltage-gated channel, shaker-related subfamily, member 8; voltage-gated potassium channel subunit Kv1.8 | |
Rabbit | |
Protein A | |
RUO | |
3744 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q16322 | |
KCNA10 | |
Synthetic peptide directed towards the middle region of human KCNA10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction