Learn More
KRT78 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 375.00 - € 515.00
Specifications
Antigen | KRT78 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18423731
|
Novus Biologicals
NBP1-93671-25ul |
25 μL |
€ 375.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18088666
|
Bio-Techne
NBP1-93671 |
0.1 mL |
€ 515.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KRT78 Polyclonal specifically detects KRT78 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KRT78 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
196374 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:VGGGLGSTCGLGSGKGSPGSCCTSIVTGGSNIILGSGKDPVLDSCSVSGSS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
CK-78, cytokeratin-78, Kb40, keratin 5b, keratin 78, keratin-78, type II cytoskeletal 78 | |
KRT78 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Product Title
KRT78 Antibody, Novus Biologicals™